Learn More
XRF Scientific Americas Inc. PLATINUM DISH SEMI-ROUND SHAPE 50ML FROM XRF SCIENTIFIC AMERICAS
Supplier:  XRF Scientific Americas Inc. DS50
Encompass
53x29mm with a 50 ml capacity. Lids sold separately. These 99.95% pure platinum dishes are available in semi-round shape with flat bottoms, and used for Fusions, Digestion, Ash testing, Loss on Ignition (LOI). Due to their high temperature strenght, ductility, corrosion and oxidation, excellent resistance to chemical attacks by acids and fusion mixtures resistance; these platinum crucibles are critical when high temperatures & resistance to chemical attacks are required.
Catalog No. NC9859311