Learn More
Sigma Aldrich Fine Chemicals Biosciences Platinum dish with reinfor1EA
Catalog No. 502909033
Encompass
Platinum labware has excellent resistance to acids such as acetic hydrochloric hydrofluoric hydroiodic nitric selenic and sulfuric and to oxidation at high temperatures. Dishes of platinum can be cleaned by boiling in a suitable solvent. For the removal of substances which have alloyed with the surface scour with alumina powder.
Catalog No. 50-290-9033 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6853211EA
Platinum labware has excellent resistance to acids such as acetic hydrochloric hydrofluoric hydroiodic nitric selenic and sulfuric and to oxidation at high temperatures. Dishes of platinum can be cleaned by boiling in a suitable solvent. For the removal of substances which have alloyed with the surface scour with alumina powder.