Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanoscience Instruments Inc PLATINUM DU NOUY RING
SDP

Catalog No. NC1638194
Encompass

  • Material: Platinum-Iridium alloy
  • Measurement method: Du Noüy ring
  • Primary application: Surface tension measurement
  • Secondary application: Interfacial tension measurement
  • Standards compliance: ISO 301, ISO 4311, ISO 6889, ASTM D1331, ASTM D 971
  • Chemical inertness: Chemically inert
  • Corrosion resistance: Highly resistant
  • Thermal stability: High
  • Wettability: Optimal
  • Cleaning: Easy to clean
  • Included accessory: Additional weight for push-mode
  • Packaging: Protective wooden box
  • Available certifications: Ring dimensions, MIKES calibration

Catalog No. NC1638194 Supplier Nanoscience Instruments Inc Supplier No. BLT106
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Material: Platinum-Iridium alloy
  • Measurement method: Du Noüy ring
  • Primary application: Surface tension measurement
  • Secondary application: Interfacial tension measurement
  • Standards compliance: ISO 301, ISO 4311, ISO 6889, ASTM D1331, ASTM D 971
  • Chemical inertness: Chemically inert
  • Corrosion resistance: Highly resistant
  • Thermal stability: High
  • Wettability: Optimal
  • Cleaning: Easy to clean
  • Included accessory: Additional weight for push-mode
  • Packaging: Protective wooden box
  • Available certifications: Ring dimensions, MIKES calibration

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.