Learn More
Sigma Aldrich Fine Chemicals Biosciences Platinum evaporating dish1EA
Catalog No. 502908877
Encompass
Platinum labware has excellent resistance to acids such as acetic hydrochloric hydrofluoric hydroiodic nitric selenic and sulfuric and to oxidation at high temperatures. Dishes of platinum can be cleaned by boiling in a suitable solvent. For the removal of substances which have alloyed with the surface scour with alumina powder.
Catalog No. 50-290-8877 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5622031EA
Platinum labware has excellent resistance to acids such as acetic hydrochloric hydrofluoric hydroiodic nitric selenic and sulfuric and to oxidation at high temperatures. Dishes of platinum can be cleaned by boiling in a suitable solvent. For the removal of substances which have alloyed with the surface scour with alumina powder.