Learn More
Strem, An Ascensus Company CAS# 7440-06-4. 25 x 25mm. Platinum foil (99.99%). MFCD00011179
SDP
Catalog No. 5090113169
Encompass
CAS# 7440-06-4. 25 x 25mm. Platinum foil (99.99%). MFCD00011179. Molecular Weight: 195.09. Molecular Formula: Pt. Color/form: 0.025mm thick (~0.34g/25x 25mm) Strem# 78-0005. http://www.strem.com/catalog/v/78-0005/
Catalog No. 50-901-13169 Supplier Strem, An Ascensus Company Supplier No. 78000525X25MM
CAS# 7440-06-4. 25 x 25mm. Platinum foil (99.99%). MFCD00011179. Molecular Weight: 195.09. Molecular Formula: Pt. Color/form: 0.025mm thick (~0.34g/25x 25mm) Strem# 78-0005. http://www.strem.com/catalog/v/78-0005/