Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum gauze, 52 mesh woven from 0.1mm (0.004in) dia wire, 99.9% (metals basis)

Catalog No. AA10283FL
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
75 x 75 mm
100 x 100 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10283FL 75 x 75 mm
AA10283FF 25 x 25 mm
AA10283FI 50 x 50 mm
AA10283GH 100 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA10283FL Supplier Thermo Scientific Chemicals Supplier No. 010283FL

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining. | CAS: 7440-06-4 | Pt | 195.08 g/mol

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Appearance Silvery white guage
Density 21.45g/cm3 at 20°C
Quantity 75 x 75 mm
Mesh Size 52 Mesh
Odor Odorless
Form Wire Cloth
Assay Percent Range 99.9%
Solubility Information Insoluble in water. Dissolves in aqua regia
Chemical Name or Material Platinum
EINECSNumber 231-116-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.