Learn More
TA Instruments Platinum HT Sample Pans 100µL Pkg of 3, for Use with All Discovery TGAs, Temperature Range Ambient to 1000°C
Catalog No. 5000580634 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Platinum HT Sample Pans 100 µL (pkg of 3) are designed for high-temperature use with all Discovery TGA instruments. They can be used over the temperature range of ambient to 1000 degrees Celsius.
Catalog No. 50-005-80634 Supplier TA Instruments Supplier No. 957571.901
Waters TA Instruments Platinum HT Sample Pans 100 µL (pkg of 3) are designed for high-temperature use with all Discovery TGA instruments. They can be used over the temperature range of ambient to 1000 degrees Celsius.