Learn More
Grainger Midwest HVAC Platinum Tool Kit, 9-Piece Set
Catalog No. 502776505
Encompass_Preferred
This HVAC platinum kit is a comprehensive set of nine essential tools designed for precision sheet metal fabrication and HVAC installation tasks. The collection features specialized snips, seamers, and folding tools to support various cutting, bending, and crimping operations, all organized within a durable tool bag for convenient transport and storage.
  • Includes left and right offset snips, bulldog snip, and left and right upright snips
  • Features a 3 in. and 6 in. interchangeable straight seamer set
  • Equipped with a 5-blade hand crimper for efficient ductwork connection
  • Contains a 12 in. folding tool and scribe marker for accurate layout and edge bending
  • Supports cutting through 18ga, 20ga, and 22ga materials
  • Supplied in a durable tool bag for protection and portability
  • Manufactured in the USA
Catalog No. 50-277-6505 Supplier Grainger Supplier No. 182N34
This HVAC platinum kit is a comprehensive set of nine essential tools designed for precision sheet metal fabrication and HVAC installation tasks. The collection features specialized snips, seamers, and folding tools to support various cutting, bending, and crimping operations, all organized within a durable tool bag for convenient transport and storage.
  • Includes left and right offset snips, bulldog snip, and left and right upright snips
  • Features a 3 in. and 6 in. interchangeable straight seamer set
  • Equipped with a 5-blade hand crimper for efficient ductwork connection
  • Contains a 12 in. folding tool and scribe marker for accurate layout and edge bending
  • Supports cutting through 18ga, 20ga, and 22ga materials
  • Supplied in a durable tool bag for protection and portability
  • Manufactured in the USA