Learn More
Strem, An Ascensus Company CAS# 10025-65-7. 1g. Platinum(II) chloride (99.9%-Pt). MFCD00011181
SDP
Catalog No. 5090113246
Encompass
CAS# 10025-65-7. 1g. Platinum(II) chloride (99.9%-Pt). MFCD00011181. Molecular Weight: 266.00. Molecular Formula: PtCl2. Color/form: olive-brown pwdr. Strem# 78-1480. http://www.strem.com/catalog/v/78-1480/
Catalog No. 50-901-13246 Supplier Strem, An Ascensus Company Supplier No. 7814801G
CAS# 10025-65-7. 1g. Platinum(II) chloride (99.9%-Pt). MFCD00011181. Molecular Weight: 266.00. Molecular Formula: PtCl2. Color/form: olive-brown pwdr. Strem# 78-1480. http://www.strem.com/catalog/v/78-1480/