Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(II) Iodide 98.5%, Thermo Scientific™

Catalog No. AC195340050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC195340050 5 g
AC195340010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC195340050 Supplier Thermo Scientific Chemicals Supplier No. 195340050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-39-8
Molecular Formula I2Pt
Molecular Weight (g/mol) 448.89
MDL Number MFCD00011183
InChI Key ZXDJCKVQKCNWEI-UHFFFAOYSA-L
Synonym platinum ii iodide, platinum iodide pti2, platinum ii iodide, premion, platinum ii iodide,premion metalsbasis ,ptmin
IUPAC Name platinum(2+) diiodide
SMILES [I-].[I-].[Pt++]

Specifications

Color Black to Gray
Physical Form Powder
Quantity 5 g
Linear Formula PtI2
Merck Index 12, 7688
Formula Weight 448.89
Percent Purity 43.0 to 44.0% (PT)
Density 6.4
Chemical Name or Material Platinum(II) iodide, 98.5%
EINECSNumber 232-204-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.