Learn More
World Precision Instrument Platinum Iridium Metal Electrodes Profile C , 305 mm long
SDP
Catalog No. 502538626
Encompass
Platinum-Iridium alloy, because of its electro chemical and mechanical characteristics, is frequently designated as the “gold standard” for chronic recording and microstimulation.
Catalog No. 50-253-8626 Supplier World Precision Instrument Supplier No. PTM123B05KT
Platinum-Iridium alloy, because of its electro chemical and mechanical characteristics, is frequently designated as the “gold standard” for chronic recording and microstimulation.