Learn More
World Precision Instrument Platinum/Iridium Wire, Teflon Coated-PTT0110
SDP
Catalog No. 50822170
Encompass
Platinum/iridium wire — uncoated (PTxxxx) and Teflon-coated (PTTxxxx) — is an alloy of 90% platinum and 10% iridium, giving excellent tensile strength and cor­ro­sion resistance.
Catalog No. 50-822-170 Supplier World Precision Instrument Supplier No. PTT0110
Platinum/iridium wire — uncoated (PTxxxx) and Teflon-coated (PTTxxxx) — is an alloy of 90% platinum and 10% iridium, giving excellent tensile strength and cor­ro­sion resistance.