Learn More
World Precision Instrument Platinum/Iridium Wire, Teflon Coated-PTT0203
SDP
Catalog No. 50822171
Encompass
Platinum/iridium wire — uncoated (PTxxxx) and Teflon-coated (PTTxxxx) — is an alloy of 90% platinum and 10% iridium, giving excellent tensile strength and cor­ro­sion resistance.
Catalog No. 50-822-171 Supplier World Precision Instrument Supplier No. PTT0203
Platinum/iridium wire — uncoated (PTxxxx) and Teflon-coated (PTTxxxx) — is an alloy of 90% platinum and 10% iridium, giving excellent tensile strength and cor­ro­sion resistance.