Learn More
World Precision Instrument Platinum/Iridium Wire for Research Applications-PT0110
SDP
Catalog No. 50822162
Encompass
Platinum/iridium wire — uncoated (PTxxxx)  is an alloy of 90% platinum and 10% iridium, giving excellent tensile strength and cor­ro­sion resistance.
Catalog No. 50-822-162 Supplier World Precision Instrument Supplier No. PT0110
Platinum/iridium wire — uncoated (PTxxxx)  is an alloy of 90% platinum and 10% iridium, giving excellent tensile strength and cor­ro­sion resistance.