Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Iridium wire, 1.0mm (0.04in) dia

Catalog No. AA42678BY
Change view
Click to view available options
Quantity:
5 cm
25 cm
100 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42678BY 100 cm
AA42678BQ 5 cm
AA42678BU 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42678BY Supplier Thermo Scientific Chemicals Supplier No. 042678BY
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Iridium wire
Name Note 1.0mm (0.04 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
Quantity 100 cm
Odor Odorless
Weight ≈0.85g/5cm
Form Wire
MDL Number MFCD02091683

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.