Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(IV) chloride, 99%

Catalog No. p-222030
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC195402500 250 mg Glass bottle
AC195400010 1 g Glass bottle
AC195400050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC195402500 Supplier Thermo Scientific Chemicals Supplier No. 195402500

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 13454-96-1 | Cl4Pt | 336.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-96-1
Molecular Formula Cl4Pt
Molecular Weight (g/mol) 336.88
MDL Number MFCD00011182
InChI Key FBEIPJNQGITEBL-UHFFFAOYSA-J
Synonym platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4
PubChem CID 26031
IUPAC Name tetrachloroplatinum
SMILES Cl[Pt](Cl)(Cl)Cl

Specifications

Melting Point 370.0°C
Color Brown
Physical Form Powder
Quantity 250 mg
Assay Percent Range 99%
Linear Formula PtCl4
Solubility Information Solubility in water: 587g/L (25°C). Other solubilities: alcohol
Formula Weight 336.89
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Platinum(IV) chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 236-645-1
RTECSNumber TP2275500
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.