Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(IV) nitrate, solution, Pt 15% w/w

Catalog No. AAH3773709
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3773709 10 g
AAH3773703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3773709 Supplier Thermo Scientific Chemicals Supplier No. H3773709
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10102-09-7 | N4O12Pt

Platinum(IV) nitrate acts as a precursor in the production of platinum dioxide on support surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-09-7
Molecular Formula N4O12Pt
MDL Number MFCD25372548
Synonym platinum 4+ ion nitric acid

Specifications

Odor Acidic
Quantity 10 g
UN Number UN3093
Solubility Information Miscible with water.
Formula Weight 443.1
Assay Pt 15% w/w
Chemical Name or Material Platinum(IV) nitrate, solution
Hazard Category H290-H314-H332
Hazard Statement GHS H Statement
H272-H314-H318-H290-H317
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
Precautionary Statement P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
EINECSNumber 432-400-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.