Learn More
Strem, An Ascensus Company ADAMS CATALYST BASFC7108 250mg
SDP
Catalog No. 5090113291
Encompass
CAS# 52785-06-5. 250mg. Platinum(IV) oxide hydrate (~80-82% Pt) (99.95+%-Pt) ADAMS' CATALYST [BASF C7018]. MFCD00066964. Molecular Weight: 227.09. Molecular Formula: PtO2∙XH2O. Color/form: brown pwdr. Strem# 78-1892. http://www.strem.com/catalog/v/78-1892/
Catalog No. 50-901-13291 Supplier Strem, An Ascensus Company Supplier No. 781892250MG
CAS# 52785-06-5. 250mg. Platinum(IV) oxide hydrate (~80-82% Pt) (99.95+%-Pt) ADAMS' CATALYST [BASF C7018]. MFCD00066964. Molecular Weight: 227.09. Molecular Formula: PtO2∙XH2O. Color/form: brown pwdr. Strem# 78-1892. http://www.strem.com/catalog/v/78-1892/