Learn More
Strem, An Ascensus Company CAS# 52785-06-5. 1g. Platinum(IV) oxide hydrate (~80-81% Pt) ADAMS' CATALYST. MFCD00066964
SDP
Catalog No. 5090113289
Encompass
CAS# 52785-06-5. 1g. Platinum(IV) oxide hydrate (~80-81% Pt) ADAMS' CATALYST. MFCD00066964. Molecular Weight: 227.09. Molecular Formula: PtO2∙XH2O. Color/form: brown pwdr. Strem# 78-1890. http://www.strem.com/catalog/v/78-1890/
Catalog No. 50-901-13289 Supplier Strem, An Ascensus Company Supplier No. 7818901G
CAS# 52785-06-5. 1g. Platinum(IV) oxide hydrate (~80-81% Pt) ADAMS' CATALYST. MFCD00066964. Molecular Weight: 227.09. Molecular Formula: PtO2∙XH2O. Color/form: brown pwdr. Strem# 78-1890. http://www.strem.com/catalog/v/78-1890/