Learn More
Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown to Black |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.8% |
| Linear Formula | PtO2·H2O |
| Fieser | 01,890 |
| Merck Index | 10, 7398 |
| Solubility Information | Solubility in water: insoluble in water. Other solubilities: soluble in conc.acids and in dil.sol. of koh |
| IUPAC Name | dioxoplatinum hydrate |
| Formula Weight | 245.1 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause fire or explosion; strong oxidizer. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.