Learn More
Electron Microscopy Sciences Platinum Loops, I.D.: 5.0 mm., 1/EA
SDP
Catalog No. 502090956
Encompass_Preferred
These platinum loops are the perfect tool for the manipulation of samples before and during sample preparation in microscopy. They are made of high purity platinum. The thickness is 0.20mm and the diameter is 5 mm ID. Comes with handle. I.D.: 5.0 mm.
Catalog No. 50-209-0956 Supplier Electron Microscopy Sciences Supplier No. 7092301
These platinum loops are the perfect tool for the manipulation of samples before and during sample preparation in microscopy. They are made of high purity platinum. The thickness is 0.20mm and the diameter is 5 mm ID. Comes with handle. I.D.: 5.0 mm.