Learn More
Glass Expansion Platinum/Nickel Hyper Skimmer Cone for PerkinElmer NexION 300/350/1000/1100/2000
SDP
Catalog No. 502702953
Encompass_Preferred
Platinum/Nickel Hyper Skimmer Cone ¿ Platinum and nickel hyper skimmer cone for PerkinElmer NexION 1000/2000/300/350 ICP-MS. Suitable for corrosive samples (e.g., high TDS samples, >5% acid matrix, HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For standard samples and routine operation, we recommend Aluminum Hyper Skimmer Cones, 501907690.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2953 Supplier Glass Expansion Supplier No. PE3015PT/NI
Platinum/Nickel Hyper Skimmer Cone ¿ Platinum and nickel hyper skimmer cone for PerkinElmer NexION 1000/2000/300/350 ICP-MS. Suitable for corrosive samples (e.g., high TDS samples, >5% acid matrix, HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For standard samples and routine operation, we recommend Aluminum Hyper Skimmer Cones, 501907690.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001