Learn More
Sigma Aldrich Platinum On Alumina Catalytic Static Mixer 316L 6Mm Dia X 150Mm
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Platinum on alumina Catalytic Static Mixer 316L 6mm dia x 150mm is a general hydrogenation/dehydrogenation catalyst that exhibits high activity with good selectivity toward carbonyl over alkene reduction. This CSM is not stable in alkaline media.
- Core: 3D printed 316L stainless steel
- Catalyst: Pt/Al2O3 wash coated
- Shape: cylindrical
- Dimensions: 6mm diameter x 150m mLengthCatalytic static mixers are a novel immobilized catalyst system, based on 3D printed mixer scaffolds containing a catalytic active layer.
A variety of different active catalysts can be deposited on the metal mixer, allowing employment in many different classes of chemical reactions, such as hydrogenations, oxidations, C-C couplings and many more.For additional information, including a general operational guide, click hereContinuous flow hydrogenations using novel catalytic static mixers inside a tubular reactorReductive aminations using a 3D printed supported metal(0) catalyst systemThe use of catalytic static mixers for continuous flow gas-liquid and transfer hydrogenations in organic synthesisCatalytic Static Mixers for the Continuous Flow Hydrogenation of a Key Intermediate of Linezolid (Zyvox)Scalable continuous flow hydrogenations using Pd/Al2O3-coated rectangular cross-section 3D-printed static mixers
Specifications
Specifications
| Molecular Formula | C32H50FN5O8 -+ xHCl |
| Linear Formula | C32H50FN5O8 -+ xHCl |
| Quantity | 1 Ea. |
| Synonym | Semi-flexible linker for PROTAC(R) development |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.