Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Platinum On Carbon

Catalog No. 33015950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-015-950G 50 g
33-015-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 33-015-950G Supplier Sigma Aldrich Supplier No. 33015950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Platinum on carbon (Pt/C) is a versatile catalyst used for the hydrogenation of alkenes, alkynes, nitro groups, ketones, and aromatics. It is used for hydrogenolysis of cyclopropanes and catalyzes the oxidation of alcohols. It is a useful reagent for reductive couplings, avoids dehalogenation.Noblyst is a registered trademark of Evonik Degussa GmbH

Specifications

Quantity 50 g
Molecular Formula Pt
Linear Formula Pt
MDL Number MFCD00011179
Synonym Pt/C
Molecular Weight (g/mol) 195.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.