Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Platinum On Carbon

Catalog No. 20593150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-593-150G 50 g
20-593-11G 1 g
20-593-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-593-150G Supplier Sigma Aldrich Supplier No. 20593150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Platinum on carbon (5wt.% loading, matrix activated carbon support) (Pt/C) can be used as a catalyst for a variety of applications, such as:hydrosilylation reaction for the production of molecules with silicon-carbon bondsdirect alcohol fuel cells (DAFCs)an electrocatalyst in hydrogen evolution reaction (HER)formation of nanocatalyst for polymer electrolyte fuel cellshydrogenation of alkenes, alkynes, nitro groups, ketones, and aromaticshydrogenolysis of cyclopropanesoxidation of alcohols.Pt/C can also be used as an hydrosilylation catalyst. It can convert alkenes and alkynes to chlorosilanes, alkylsilanes, and alkoxysilanes at the temperature and pressure conditions of 100-300 oC and 30-1000 psi, respectively.It may be also used as a catalyst in methanol fuel cells.

Specifications

Quantity 50 g
Molecular Formula Pt
Linear Formula Pt
MDL Number MFCD00011179
Synonym Pt/C
Molecular Weight (g/mol) 195.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.