Learn More
Protech International Inc Platinum Plate Petridish Electrode, 1mm Gap x 15mm Length
SDP
Catalog No. NC1879373
Encompass
Platinum plate electrode on tempered glass petridish, can hold up to ~80 mouse zygotes in a 8 L electroporation buffer. This enables efficient and high throughput zygote genome editing using CRISPR/Cas9 by electroporation.
Catalog No. NC1879373 Supplier Protech International Inc Supplier No. PD1.15
 Restricted Item
Add to Cart
Add to Cart
Platinum plate electrode on tempered glass petridish, can hold up to ~80 mouse zygotes in a 8 L electroporation buffer. This enables efficient and high throughput zygote genome editing using CRISPR/Cas9 by electroporation.