Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Potassium Chloride (Laboratory), Fisher Chemical™

Catalog No. P1581 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P158-1 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P158-1

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16921-30-5
Molecular Formula Cl6K2Pt
Molecular Weight (g/mol) 485.98
MDL Number MFCD00011389
InChI Key DPAIVKJGTXERIM-UHFFFAOYSA-H
Synonym potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride
PubChem CID 61856
IUPAC Name dipotassium hexachloroplatinumdiuide
SMILES [K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 250°C
Color Yellow to Orange
Physical Form Powder/Solid
Quantity 1 g
UN Number 3288
Formula Weight 486.01
Grade Laboratory
Identification Pass Test
Packaging Glass Bottle
Density 3.499g/cm³
Chemical Name or Material Platinum Potassium Chloride
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.