Learn More
Strem, An Ascensus Company CAS# 7440-06-4. 5g. Platinum powder (99.9%). MFCD00011179
SDP
Catalog No. 502016656
Encompass
CAS# 7440-06-4. 5g. Platinum powder (99.9%). MFCD00011179. Molecular Weight: 195.09. Molecular Formula: Pt. Color/form: avg. particle size 0.27-0.47 micron Strem# 93-7833. http://www.strem.com/catalog/v/93-7833/
Catalog No. 50-201-6656 Supplier Strem, An Ascensus Company Supplier No. 9378335G
CAS# 7440-06-4. 5g. Platinum powder (99.9%). MFCD00011179. Molecular Weight: 195.09. Molecular Formula: Pt. Color/form: avg. particle size 0.27-0.47 micron Strem# 93-7833. http://www.strem.com/catalog/v/93-7833/