Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Platinum-ruthenium alloy on graphitized carbon

Catalog No. 7385731G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g

Catalog No. 73-857-31G

Supplier: Sigma Aldrich 7385731G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Platinum-ruthenium alloy on graphitized carbon (30% Pt-Ru/Vulcan) can be used as an electrocatalyst for the fabrication of direct methanol fuel cells (DMFC) and proton exchange membrane (PEM) based fuel cells.We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Specifications

Linear Formula Pt/Ru
Quantity 1 g
MDL Number MFCD00798552
Synonym 20% Pt, 10% Ru on Vulcan XC72; 30% Pt-Ru/Vulcan
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.