Learn More
TA Instruments Platinum Sample Pans 100µL Pkg of 3, for Use with Discovery SA and All Discovery TGAs, Temperature Range Ambient to 1000°C
Catalog No. 5000580633 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Platinum Sample Pans 100 µL (pkg of 3) are designed for use with Discovery SA and all Discovery TGA instruments. They can be used over the temperature range of ambient to 1000 degrees Celsius.
Catalog No. 50-005-80633 Supplier TA Instruments Supplier No. 957207.904
Waters TA Instruments Platinum Sample Pans 100 µL (pkg of 3) are designed for use with Discovery SA and all Discovery TGA instruments. They can be used over the temperature range of ambient to 1000 degrees Celsius.