Learn More
TA Instruments Platinum Sample Pans 50µL Pkg of 3, for Use with Discovery SA and All Discovery TGAs, Temperature Range Ambient to 1000°C
Catalog No. 5000580632 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Platinum Sample Pans 50 µL (pkg of 3) are designed for use with Discovery SA and all Discovery TGA instruments. They can be used over the temperature range of ambient to 1000 degrees Celsius.
Catalog No. 50-005-80632 Supplier TA Instruments Supplier No. 957207.903
Waters TA Instruments Platinum Sample Pans 50 µL (pkg of 3) are designed for use with Discovery SA and all Discovery TGA instruments. They can be used over the temperature range of ambient to 1000 degrees Celsius.