Learn More
TA Instruments Platinum Sample Pans, DSC Platinum Pans, Experiments up to 725°C, High Thermal Stability, Inert for Accurate DSC Measurements, 10 Pack
Catalog No. 5000580883 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Platinum Sample Pans are DSC platinum pans useful for experiments up to 725°C, providing high thermal stability and inertness for precise high-temperature DSC measurements.
Catalog No. 50-005-80883 Supplier TA Instruments Supplier No. 900578.901
Waters TA Instruments Platinum Sample Pans are DSC platinum pans useful for experiments up to 725°C, providing high thermal stability and inertness for precise high-temperature DSC measurements.