Learn More
Glass Expansion Platinum Skimmer Cone for Agilent 4500/7500, Suitable for: Agilent 4500, 7500a (G1820-65237). Must be mounted on the brass skimmer base.
SDP
Catalog No. 501907573
Encompass_Preferred
Platinum Skimmer Cone (G1820-65237) - For Agilent 4500/7500s/a/i. Recommended for corrosive acids, including HF. For use with Pt sampling cone, and must be mounted on the copper skimmer base. Platinum cones are not for use with T-mode specifications with 7500s/a/i. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1002) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, we recommend the standard Nickel Skimmer, see catalog number 501907566.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7573 Supplier Glass Expansion Supplier No. AT1008PT
Platinum Skimmer Cone (G1820-65237) - For Agilent 4500/7500s/a/i. Recommended for corrosive acids, including HF. For use with Pt sampling cone, and must be mounted on the copper skimmer base. Platinum cones are not for use with T-mode specifications with 7500s/a/i. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1002) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, we recommend the standard Nickel Skimmer, see catalog number 501907566.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001