Learn More
Glass Expansion Platinum Sampler Cone for Agilent 7700/7800/7900/8800/8900, 18mm insert (G3280-67056)
SDP
Catalog No. 501907582
Encompass_Preferred
Platinum Sampler Cone with 18 mm insert (G3280-67056) ¿ For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. For use with high viscosity or high boiling point acids (e.g., H2SO4, H3PO4). Best LOD in high matrix samples. Use with platinum skimmer cone and brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Sampler with Ni Base, P/N 501907586. For high TDS samples causing rapid cone deposition, use Pt Sampler with Ni Base, P/N 501907585. For lowest Ni background & laser ablation applications, use Al Sampler, see AT7701-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7582 Supplier Glass Expansion Supplier No. AT7706APT
Add to Cart
Add to Cart
Request a Quote
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Platinum Sampler Cone with 18 mm insert (G3280-67056) ¿ For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. For use with high viscosity or high boiling point acids (e.g., H2SO4, H3PO4). Best LOD in high matrix samples. Use with platinum skimmer cone and brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Sampler with Ni Base, P/N 501907586. For high TDS samples causing rapid cone deposition, use Pt Sampler with Ni Base, P/N 501907585. For lowest Ni background & laser ablation applications, use Al Sampler, see AT7701-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001