Learn More
Glass Expansion Platinum Skimmer Cone with Copper Base for s-lens (G3280-67064) Suitable for Agilent 7700s/7900
SDP
Catalog No. 501907588
Encompass_Preferred
Platinum Skimmer Cone with Copper Base for s-lens (G3280-67064) ¿ For Agilent 7700s/7900/8800 ICP-MS. Standard cone for use with s-lens, and must be mounted on the brass skimmer adaptor base. Recommended for corrosive acids, including HF and semiconductor samples. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, we recommend the standard Nickel Skimmer for s-lens, see catalog number 501907578. For organic analysis, use Pt Skimmer with Ni Base for s-lens, P/N 501907589, which must be mounted on brass adaptor skimmer base.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7588 Supplier Glass Expansion Supplier No. AT7708SPT
Platinum Skimmer Cone with Copper Base for s-lens (G3280-67064) ¿ For Agilent 7700s/7900/8800 ICP-MS. Standard cone for use with s-lens, and must be mounted on the brass skimmer adaptor base. Recommended for corrosive acids, including HF and semiconductor samples. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, we recommend the standard Nickel Skimmer for s-lens, see catalog number 501907578. For organic analysis, use Pt Skimmer with Ni Base for s-lens, P/N 501907589, which must be mounted on brass adaptor skimmer base.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001