Learn More
Glass Expansion Platinum Sampler Cone for PerkinElmer NexION 300/350/1000/1100/2000/2200/5000 ICP-MS (W1033614)
SDP
Catalog No. 501907686
Encompass_Preferred
Platinum Sampler Cone (W1033614) ¿ For PerkinElmer NexION 1000/2000/5000/300/350 ICP-MS. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Replace Sampler Cone Gasket each time the cone is installed, even if it is the same cone (501907691). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use Sampler ConeGuard to protect cone threads during cleaning, 70-803-1024.For most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic) we recommend Nickel Sampler Cones, see catalog number 501907684. For lowest boron background, we recommend Boron-Free Platinum Sampler Cones, see catalog number 501907687.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7686 Supplier Glass Expansion Supplier No. PE3013PT
Platinum Sampler Cone (W1033614) ¿ For PerkinElmer NexION 1000/2000/5000/300/350 ICP-MS. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Replace Sampler Cone Gasket each time the cone is installed, even if it is the same cone (501907691). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use Sampler ConeGuard to protect cone threads during cleaning, 70-803-1024.For most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic) we recommend Nickel Sampler Cones, see catalog number 501907684. For lowest boron background, we recommend Boron-Free Platinum Sampler Cones, see catalog number 501907687.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001