Learn More
Glass Expansion Platinum Sampler Cone for PerkinElmer Elan 6000/9000/DRCII/DRC-E (WE027802)
SDP
Catalog No. 501907682
Encompass_Preferred
Platinum Sampler Cone (WE027802) ¿ For PerkinElmer Elan 6000/9000/DRCII/DRC-E ICP-MS. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Replace sampler cone O-ring if brittle, cracked or damaged (NC1585741). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For routine samples, we recommend Nickel Sampler Cones, see catalog number NC1585739.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7682 Supplier Glass Expansion Supplier No. PE2013PT
Platinum Sampler Cone (WE027802) ¿ For PerkinElmer Elan 6000/9000/DRCII/DRC-E ICP-MS. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Replace sampler cone O-ring if brittle, cracked or damaged (NC1585741). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For routine samples, we recommend Nickel Sampler Cones, see catalog number NC1585739.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001