Learn More
Glass Expansion Platinum Sampler Cone, Wet Plasma (319-593)
SDP
Catalog No. 501907678
Encompass_Preferred
Platinum Sampler Cone, Wet Plasma (319-593) For Nu Instruments Plasma2, Plasma3, Attom ES ICP-MS. Suitable for standard applications. Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7678 Supplier Glass Expansion Supplier No. NU1006APT
Platinum Sampler Cone, Wet Plasma (319-593) For Nu Instruments Plasma2, Plasma3, Attom ES ICP-MS. Suitable for standard applications. Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001