Learn More
Glass Expansion Platinum Sampler Cone (Boron-Free) for PerkinElmer NexION 300/350/1000/1100/2000/2200/5000 ICP-MS
SDP
Catalog No. 501907687
Encompass_Preferred
Platinum Sampler Cone, Boron-Free ¿ For PerkinElmer NexION 1000/2000/5000/300/350 ICP-MS. Suitable for applications requiring low boron background, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Replace Sampler Cone Gasket each time the cone is installed, even if it is the same cone (501907691). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use Sampler ConeGuard to protect cone threads during cleaning, 70-803-1024.For most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic) we recommend Nickel Sampler Cones, see catalog number 501907684. For corrosive samples use Platinum Sampler Cones, see catalog number 501907686.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7687 Supplier Glass Expansion Supplier No. PE3013PTBF
Platinum Sampler Cone, Boron-Free ¿ For PerkinElmer NexION 1000/2000/5000/300/350 ICP-MS. Suitable for applications requiring low boron background, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Replace Sampler Cone Gasket each time the cone is installed, even if it is the same cone (501907691). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use Sampler ConeGuard to protect cone threads during cleaning, 70-803-1024.For most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic) we recommend Nickel Sampler Cones, see catalog number 501907684. For corrosive samples use Platinum Sampler Cones, see catalog number 501907686.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001