Learn More
Glass Expansion Platinum Sampler Cone for Agilent 7700/7800/7850/7900/8800/8900 ICP-MS (G3280-67036)
SDP
Catalog No. 501907584
Encompass_Preferred
Platinum Sampler Cone (G3280-67036) ¿ For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. Standard with s-lens for semiconductor applications. Recommended for high TDS samples and for lower detection limits. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Use with platinum skimmer cone and brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Sampler with Ni Base, P/N 501907586. For high TDS samples causing rapid cone deposition, use Pt Sampler with Ni Base, P/N 501907585. For lowest Ni background & laser ablation applications, use Al Sampler, see AT7701-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7584 Supplier Glass Expansion Supplier No. AT7706PT
Platinum Sampler Cone (G3280-67036) ¿ For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. Standard with s-lens for semiconductor applications. Recommended for high TDS samples and for lower detection limits. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Use with platinum skimmer cone and brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Sampler with Ni Base, P/N 501907586. For high TDS samples causing rapid cone deposition, use Pt Sampler with Ni Base, P/N 501907585. For lowest Ni background & laser ablation applications, use Al Sampler, see AT7701-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001