Learn More
Glass Expansion Platinum Skimmer Cone, Copper Base - 7500cs, 7500ce, Suitable for: Agilent 7500cs, 7500ce (G1833-65132). Must be mounted on the brass skimmer base.
SDP
Catalog No. 501907571
Encompass_Preferred
Platinum Skimmer Cone, Copper Base (G1833-65132) - For Agilent 7500cs/7500ce. Recommended for corrosive acids, including HF. For use with Pt sampling cone, and must be mounted on the brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, we recommend the standard Nickel Skimmer, see catalog number 501907564. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7571 Supplier Glass Expansion Supplier No. AT1008CSPT
Platinum Skimmer Cone, Copper Base (G1833-65132) - For Agilent 7500cs/7500ce. Recommended for corrosive acids, including HF. For use with Pt sampling cone, and must be mounted on the brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, we recommend the standard Nickel Skimmer, see catalog number 501907564. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001