Learn More
Glass Expansion Platinum Sampler Cone with Nickel Base for Agilent 7700/7800/7900/8800/8900 (15mm insert)
SDP
Catalog No. 501907583
Encompass_Preferred
Platinum Sampler Cone (15 mm insert) with Nickel Base – For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. For use with aggressive acid matrices (e.g., HCl, HF, HClO4) and high matrix samples when cone deposition is rapid due to high TDS. Use with platinum skimmer cone and brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For routine samples with x-lens, use P/N 501907575. For high viscosity or high boiling point acids (e.g., H2SO4, H3PO4), use Pt Sampler with 18 mm insert, see P/N 501907582. For lowest Ni background & laser ablation applications, use Al Sampler, see AT7701-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7583 Supplier Glass Expansion Supplier No. AT7706BPT/NI
Add to Cart
Add to Cart
Request a Quote
Platinum Sampler Cone (15 mm insert) with Nickel Base – For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. For use with aggressive acid matrices (e.g., HCl, HF, HClO4) and high matrix samples when cone deposition is rapid due to high TDS. Use with platinum skimmer cone and brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For routine samples with x-lens, use P/N 501907575. For high viscosity or high boiling point acids (e.g., H2SO4, H3PO4), use Pt Sampler with 18 mm insert, see P/N 501907582. For lowest Ni background & laser ablation applications, use Al Sampler, see AT7701-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001