Learn More
Perkin Elmer US LLC Platinum Sampler Cone for Sulfur Based Acids/Solvents (18 mm) for NexION 300/350/1000/2000/5000
Catalog No. 502104223
Encompass_Preferred
This platinum sampler cone is used with NexION 300, 350, 1000, 2000, and 5000 ICP-MS series instruments.
Catalog No. 50-210-4223 Supplier Perkin Elmer US LLC Supplier No. N8145028
Add to Cart
Add to Cart
Request a Quote
This platinum sampler cone is used with NexION 300, 350, 1000, 2000, and 5000 ICP-MS series instruments.