Learn More
Glass Expansion Platinum Sampler Ni Plated
SDP
Catalog No. 502703002
Encompass_Preferred
Platinum Sampler Cone with Nickel Plated Base (Extended Life) – For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. Ideal for when cone deposition is rapid due to high TDS and more durability is required. For use with aggressive acid matrices (e.g., HCl, HF, HClO4) and high matrix samples. Use with platinum skimmer cone and brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For routine samples with x-lens, use P/N 501907575. For high TDS samples causing rapid cone deposition, use Pt Sampler with Ni Base, P/N 501907585 or 501907583. For lowest Ni background & laser ablation applications, use Al Sampler, see AT7701-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3002 Supplier Glass Expansion Supplier No. AT7706LPT/NIP
Platinum Sampler Cone with Nickel Plated Base (Extended Life) – For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. Ideal for when cone deposition is rapid due to high TDS and more durability is required. For use with aggressive acid matrices (e.g., HCl, HF, HClO4) and high matrix samples. Use with platinum skimmer cone and brass skimmer base. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For routine samples with x-lens, use P/N 501907575. For high TDS samples causing rapid cone deposition, use Pt Sampler with Ni Base, P/N 501907585 or 501907583. For lowest Ni background & laser ablation applications, use Al Sampler, see AT7701-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001