Learn More
Hillrom Platinum Series DS58 Hand Aneroids
Catalog No. 22355145
Change view
Click to view available options
For Use With (Equipment):
Blood Pressure Cuff
Catalog No. For Use With (Equipment)
22-355-145 Blood Pressure Cuff
1 options
This item is not returnable. View return policy
Catalog No. 22-355-145 Supplier Hillrom Supplier No. WEL DS58
Add to Cart
Add to Cart
This item is not returnable. View return policy
With gear-free DuraShock™ technology, you can count on the DS58 aneroid to remain in calibration longer than traditional aneroids, providing more accurate readings over time and a lower lifetime cost, helping to improve practice efficiencies and your bottom line.
  • Gear-free DuraShock™ technology
  • Certified accuracy to ±3 mmHg
  • Laser-engraved dial face for accuracy
  • Jeweled movement for long life
  • Lifetime calibration warranty
  • Latex-free for safety
  • Thumbscrew bleed valve and ergonomic inflation bulb
  • Unsurpassed reliability in a traditional, handheld design
  • Designed to enhance comfort
  • Withstands a 60 in. drop, exceeds AAMI's shock resistant requirement
Specifications
For Use With (Equipment) Blood Pressure Cuff
Includes Lifetime Calibration Warranty
Product Type Gauge