Learn More
Glass Expansion Platinum Skimmer Cone with Copper Base for Agilent 8900 with S lens (G3666-67401)
SDP
Supplier:  Glass Expansion AT8908SPT/CU
Encompass_Preferred
Platinum Skimmer Cone with Copper Base for s-lens (G3666-67401) ¿ For Agilent 8900 ICP-MS. Recommended for corrosive samples, including HF (e.g., routine aqueous environmental samples, >5% acid matrix, HF). More durable, longer-lasting, & because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, use standard Nickel Skimmer for x-lens, see P/N 501907593.For organic analysis, use Platinum Skimmer with Ni Base for x-lens, P/N 501907595, which must be mounted on brass adaptor skimmer base, see P/N AT7905X-BR. For lowest nickel background and laser ablation applications, use Aluminum Skimmer for x-lens, see P/N AT7902X-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7596
 Restricted Item