Learn More
Glass Expansion Platinum Skimmer Cone (H, with boron), Suitable for: Thermo/Finnigan (105197)
SDP
Catalog No. 501907716
Encompass_Preferred
Platinum Skimmer Cone, H, with Boron (1047461) ¿ For Thermo/Finnigan Element 1, 2 & Neptune ICP-MS. Ideal for semiconductor applications. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest copper background, we reccommend Nickel Skimmer, see catalog number 501907706.For high sensitivity applications we reccommend Nickel X-Skimmer with Copper Core, see catalog number 501907707.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7716 Supplier Glass Expansion Supplier No. FL9001NI
Platinum Skimmer Cone, H, with Boron (1047461) ¿ For Thermo/Finnigan Element 1, 2 & Neptune ICP-MS. Ideal for semiconductor applications. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest copper background, we reccommend Nickel Skimmer, see catalog number 501907706.For high sensitivity applications we reccommend Nickel X-Skimmer with Copper Core, see catalog number 501907707.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001