Learn More
Perkin Elmer US LLC Platinum Skimmer Cone for NexION 300/350/1000/2000
Catalog No. NC0336288
Encompass_Preferred
Precision-designed and manufactured, large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material for the majority of sample types, while platinum is the material of choice for more corrosive samples. They have been designed to maximize signal stability and to minimize clogging during extended runs of samples containing high dissolved solids.
Catalog No. NC0336288 Supplier Perkin Elmer US LLC Supplier No. W1026907
 Restricted Item
Add to Cart
Add to Cart
Precision-designed and manufactured, large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material for the majority of sample types, while platinum is the material of choice for more corrosive samples. They have been designed to maximize signal stability and to minimize clogging during extended runs of samples containing high dissolved solids.