Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collibri™ Library Amplification Master Mix with Primer Mix

Catalog No. A38540050
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
50
250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A38540050 50
A38540250 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A38540050 Supplier Invitrogen™ Supplier No. A38540050
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen™ Platinum SuperFi Library Amplification Master Mix is a ready-to-use solution designed for amplification of next-generation sequencing (NGS) libraries.

Master mix includes Platinum SuperFi™ DNA Polymerase in combination with a proprietary reaction buffer containing all necessary components for efficient library amplification with minimal bias and superior accuracy.

 Features include:

  • High fidelity—minimal error rates similar to PCR-free methods
  • High efficiency—high yields from minute input amounts
  • Minimal bias—uniform amplification regardless of GC content

 

  • Supplied with a 10X Primer Mix that targets the P5 and P7 regions of Illumina adapters. The master mix is supplemented with an inert blue dye, while the primer mix contains a yellow dye. Mixing both components in a PCR reaction turns the final PCR solution green. This provides a visual aid when pipetting and helps decrease the risk of pipetting errors during reaction setup
  • Platinum SuperFi™ DNA Polymerase has both 5’ to 3’ polymerase and 3’ to 5’ exonuclease (proofreading) activities, but lacks 5’ to 3’ exonuclease activity. Exceptionally strong proofreading activity ensures amplification of NGS libraries with supreme sequence accuracy, producing NGS error rates similar to PCR-free methods
  • Proprietary reaction buffer contains all necessary PCR components and additives to enable efficient and uniform library amplification regardless of GC content, helping improve coverage across GC- and AT-rich sequences and other complex regions. High amplification efficiency enables the use of fewer cycles to achieve the desired yield
  • Platinum hot-start technology inhibits DNA polymerase activity at ambient temperatures, allowing room temperature reaction setup and storage of pre-assembled PCR reactions for up to 24 hours prior to the PCR.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 x 1.25 mL Collibri Library Amplification Master Mix (2X)
• 1 x 500 µL Primer Mix

Store at -20°C.
Sample Type DNA, RNA
Fidelity (vs. Taq) ≥100
GC-Rich PCR Performance High
For Use With (Application) Sequencing
Hot Start Built-In Hot Start
No. of Reactions 50
Overhang Blunt
Product Line Collibri
Product Type Library Amplification Master Mix
Quantity 50 reactions
Reaction Format SuperMix or Master Mix
Shipping Condition Dry Ice
Test Time Standard
Form Liquid
Show More Show Less
Are PCR products generated using the Collibri Library Amplification Master Mix blunt-ended?

Yes, PCR products generated using the Collibri Library Amplification Master Mix are blunt-ended.

Is Collibri Library Amplification Master Mix compatible with magnetic beads?

Yes, Collibri Library Amplification Master Mix is compatible with magnetic beads provided in Collibri library preparation kits and in Agencourt AmPure XP system from Beckman Coulter.

What is the fidelity of the Collibri Library Amplification Master Mix?

The Collibri Library Amplification Master Mix contains Platinum SuperFi DNA Polymerase that has strong 3' to 5' exonuclease (proofreading) activity for superior amplification accuracy, greater than 100-fold in comparison with Taq polymerase.

What are the recommended storage conditions for the Collibri Library Amplification Master Mix?

We recommend storing the Collibri Library Amplification Master Mix at -20 degrees C.

Is Collibri Library Amplification Master Mix suitable for amplification of libraries prepared from FFPE samples?

Yes, however, additional PCR cycles (1-3) are typically needed with FFPE DNA or other challenging samples. To determine the optimal PCR conditions, use the table in the user manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017582_Platinum_SuperFi_LibraryAmp_MM_w_Primer_Mix_UG.pdf) with the required number of PCR cycles per sample size as a starting point.

How many PCR cycles should I use for library amplification with the Collibri Library Amplification Master Mix?

To determine the optimal PCR conditions, use the table in the user manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017582_Platinum_SuperFi_LibraryAmp_MM_w_Primer_Mix_UG.pdf) with the required number of PCR cycles per sample size as a starting point. Add additional PCR cycles to amplify libraries prepared from FFPE DNA or other challenging samples.

How should I determine the optimal annealing temperature to use with the Collibri Library Amplification Master Mix?

An annealing temperature of 60 degrees C is recommended for the Primer Mix supplied with the kit (Cat. No. A38540050, A38540250). Optimization of annealing temperature may be required for different user-supplied adaptors and primers. Note that the annealing temperature for Platinum SuperFi DNA Polymerase is typically higher than that for other DNA polymerases. To determine the optimal annealing conditions, use the Tm calculator on our website at thermofisher.com/tmcalculator as a starting point. If necessary, use a temperature gradient to optimize further and empirically determine the ideal annealing temperature. The annealing gradient should extend up to the extension temperature (two-step PCR).

Is Collibri Library Amplification Master Mix compatible with bisulfite-treated DNA?

No. Collibri Library Amplification Master Mix contains Platinum SuperFi DNA Polymerase that does not incorporate dUTP and is not compatible with uracil-containing primers and templates. For this reason, Collibri Library Amplification Master Mix is not suitable for amplification of bisulfite-converted DNA.

Does Collibri Library Amplification Master Mix incorporate dUTP?

No. Collibri Library Amplification Master Mix contains Platinum SuperFi DNA Polymerase that does not incorporate dUTP and is not compatible with uracil-containing primers and templates. For this reason, Collibri Library Amplification Master Mix is not suitable for amplification of bisulfite-converted DNA.

Is Collibri Library Amplification Master Mix compatible with AmpliSeq technology?

No, this product is not designed for amplification of AmpliSeq libraries.

What is the difference between Platinum SuperFi PCR Master Mix and Collibri Library Amplification Master Mix?

Both master mixes include Platinum SuperFi DNA Polymerase that has strong 3' to 5' exonuclease (proofreading) activity for superior amplification accuracy. However, the master mix composition is different. Platinum SuperFi PCR Master Mix has been optimized for end-point PCR amplification of a variety of templates, including long and difficult samples. Collibri Library Amplification Master Mix is based on a proprietary reaction buffer that has been specifically optimized for efficient and uniform amplification of NGS libraries regardless of GC content.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.