Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ SuperFi™ U Multiplex Master Mix

Catalog No. A5140096
Encompass_Preferred
Change view
Click to view available options
No. of Samples:
96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Samples
A5140096 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5140096 Supplier Invitrogen™ Supplier No. A5140096
Add to Cart
Add to Cart

Invitrogen Platinum SuperFi U Multiplex Master Mix is a product designed to synthesize multiple custom amplicons for further use in next-generation sequencing.

  • Designed to synthesize multiple custom amplicons for further use in next-generation sequencing
  • The ready-to-use 4X master mix contains uracil-tolerant high-fidelity DNA polymerase with all necessary buffer components and dNTPs for efficient targeted amplification
  • Optimized for high multiplex PCR
  • Enables fast, efficient, and robust amplification
  • Provides high yields
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Concentration 4X
Content And Storage Store at -25°C to -15°C
Sample Type gDNA, cDNA
Enzyme Platinum™ SuperFi™ U DNA Polymerase
For Use With (Application) Sars-Covid-2 Sequencing
No. of Reactions 96 Preps
No. of Samples 96 preps
Product Line Collibri
Product Type Master Mix
Quantity 96 reactions
Species All
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.